Nicotinic acid (Niacin), Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3-Pyridinecarboxylic acid, Pyridine-3-carboxylic acid, 3-Picolinic acid, Niacin, Nicotinic acid, Vitamin B3
72309-100G 72309-500G
SIAL72309-100GEA 50.4 EUR
SIAL72309-100G SIAL72309-500G
Nicotinic acid (Niacin), Sigma-Aldrich®
Nicotinic acid (Niacin)
Formula: C₆H₅NO₂
MW: 123.11 g/mol
Boiling Pt: 258
Melting Pt: 142
Density: 1.473 g/cm³ (20 °C)
Flash Pt: 193
Storage Temperature: Ambient
MDL Number: MFCD00006391
CAS Number: 59-67-6
EINECS: 200-441-0
Merck Index: 13,06552