Isonicotinic acid
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Pyridine-4-carboxylic acid, 4-Pyridinecarboxylic acid, 4-Picolinic acid
CC04201.1
CC04201.10
MAYBCC04201.1EA
0
EUR
MAYBCC04201.1
MAYBCC04201.10
Isonicotinic acid
Isonicotinic acid
Formula:
C₆H₅NO₂ MW: 123.11 g/mol Boiling Pt: 396 °C (1013 hPa) Melting Pt: 315…319 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006429 CAS Number: 55-22-1 EINECS: 200-228-2 Merck Index: 13,05206 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...