6-Hydroxynicotinic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
6-Oxo-1,6-dihydropyridine-3-carboxylic acid, 6-Hydroxy-3-pyridinecarboxylic acid, 6-Hydroxyniacin, 6-Hydroxypyridine-3-carboxylic acid
M-Clarity™ quality level = MQ100
128759-100G
128759-25G
SIAL128759-100GEA
190
EUR
SIAL128759-100G
SIAL128759-25G
6-Hydroxynicotinic acid, Sigma-Aldrich®
6-Hydroxynicotinic acid
Formula:
C₆H₅NO₃ MW: 139.11 g/mol Melting Pt: 304…306 °C |
MDL Number:
MFCD00006277 CAS Number: 5006-66-6 EINECS: 225-682-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...