Pyridine-2,6-dicarboxylic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2,6-Pyridinedicarboxylic acid, Dipicolinic acid, DPA
M-Clarity™ quality level = MQ200
P63808-100G P63808-25G P63808-500G
SIALP63808-100GEA 58.4 EUR
SIALP63808-100G SIALP63808-25G SIALP63808-500G
Pyridine-2,6-dicarboxylic acid, Sigma-Aldrich®
Pyridine-2,6-dicarboxylic acid
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 357
Melting Pt: 327
Flash Pt: 188
Storage Temperature: Ambient
MDL Number: MFCD00006299
CAS Number: 499-83-2
EINECS: 207-894-3