2-Fluoro-5-iodopyridine, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
660043-10G
660043-1G
SIAL660043-10GEA
174
EUR
SIAL660043-10G
SIAL660043-1G
2-Fluoro-5-iodopyridine, Sigma-Aldrich®
2-Fluoro-5-iodopyridine
Formula:
C₅H₃FIN MW: 222.99 g/mol |
MDL Number:
MFCD03095301 CAS Number: 171197-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...