4-(Dimethylamino)pyridine, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 4-Dimethylaminopyridine, DMAP, N,N-Dimethylpyridin-4-amine

M-Clarity™ quality level = MQ200

107700-25G 107700-100G 107700-5G
SIAL107700-25GEA 60 EUR
SIAL107700-25G SIAL107700-100G SIAL107700-5G
4-(Dimethylamino)pyridine, Sigma-Aldrich®
4-(Dimethylamino)pyridine
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 195
Melting Pt: 109…111 °C
Flash Pt: 124
MDL Number: MFCD00006418
CAS Number: 1122-58-3
EINECS: 214-353-5
UN: 2811
ADR: 6.1 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR