4-(Dimethylamino)pyridine, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
4-Dimethylaminopyridine, DMAP, N,N-Dimethylpyridin-4-amine
M-Clarity™ quality level = MQ200
39405-10G
39405-1KG
39405-250G
39405-50G
SIAL39405-10GEA
55.5
EUR
SIAL39405-10G
SIAL39405-1KG
SIAL39405-250G
SIAL39405-50G
4-(Dimethylamino)pyridine, Sigma-Aldrich®
4-(Dimethylamino)pyridine
Formula:
C₇H₁₀N₂ MW: 122.17 g/mol Boiling Pt: 195 Melting Pt: 109…111 °C Flash Pt: 124 |
MDL Number:
MFCD00006418 CAS Number: 1122-58-3 EINECS: 214-353-5 UN: 2811 ADR: 6.1 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...