2-Aminopyridine, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Pyridylamine, Pyridin-2-amine
M-Clarity™ quality level = MQ100
A77989-100G A77989-25G
SIALA77989-100GEA 110 EUR
SIALA77989-100G SIALA77989-25G
2-Aminopyridine, Sigma-Aldrich®
2-Aminopyridine
Formula: C₅H₆N₂
MW: 94.12 g/mol
Boiling Pt: 210.6
Melting Pt: 50…52 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 92
Storage Temperature: Ambient
MDL Number: MFCD00006312
CAS Number: 504-29-0
EINECS: 207-988-4
UN: 2671
ADR: 6.1 II
Merck Index: 13,00472