3-Aminopyridine, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Pyridin-3-amine, 3-Pyridylamine
A78209-100G
A78209-25G
A78209-500G
SIALA78209-100GEA
125
EUR
SIALA78209-100G
SIALA78209-25G
SIALA78209-500G
3-Aminopyridine, Sigma-Aldrich®
3-Aminopyridine
Formula:
C₅H₆N₂ MW: 94.12 g/mol Boiling Pt: ∼248 °C (1013 hPa) Melting Pt: 60…63 °C Flash Pt: 88 Storage Temperature: Ambient |
MDL Number:
MFCD00006400 CAS Number: 462-08-8 EINECS: 207-322-2 UN: 2671 ADR: 6.1 II Merck Index: 13,00472 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...