4-Aminopyridine, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Pyridin-4-amine, 4-Pyridylamine
M-Clarity™ quality level = MQ200
A78403-25G
A78403-100G
SIALA78403-25GEA
50.2
EUR
SIALA78403-25G
SIALA78403-100G
4-Aminopyridine, Sigma-Aldrich®
4-Aminopyridine
Formula:
C₅H₆N₂ MW: 94.12 g/mol Boiling Pt: 273 Melting Pt: 157…160 °C Density: 1.25 g/cm³ (20 °C) Flash Pt: 156 Storage Temperature: Ambient |
MDL Number:
MFCD00006439 CAS Number: 504-24-5 EINECS: 207-987-9 UN: 2671 ADR: 6.1 II Merck Index: 13,03964 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...