2-Amino-3-hydroxypyridin, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
2-Aminopyridin-3-ol, 2-Amino-3-hydroxypyridine
M-Clarity™ quality level = MQ100
122513-100G
122513-25G
SIAL122513-100GEA
361
EUR
SIAL122513-100G
SIAL122513-25G
2-Amino-3-hydroxypyridin, Sigma-Aldrich®
2-Amino-3-hydroxypyridin
Formula:
C₅H₆N₂O MW: 110.12 g/mol Boiling Pt: 250 °C (1013 hPa) Melting Pt: 172 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006317 CAS Number: 16867-03-1 EINECS: 240-886-8 UN: 2811 ADR: 6.1,III |