2,3-Diaminopyridine, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Pyridine-2,3-diyldiamine
M-Clarity™ quality level = MQ100
125857-25G
125857-5G
SIAL125857-25GEA
537
EUR
SIAL125857-25G
SIAL125857-5G
2,3-Diaminopyridine, Sigma-Aldrich®
2,3-Diaminopyridine
Formula:
C₅H₇N₃ MW: 109.13 g/mol Boiling Pt: 148…150 °C (5 mmHg) Melting Pt: 110…112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006319 CAS Number: 452-58-4 EINECS: 207-200-9 UN: 2671 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...