2,5-Diaminopyridine, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Pyridine-2,5-diamine, Pyridine-2,5-diyldiamine
M-Clarity™ quality level = MQ100
530603-10G
530603-1G
530603-5G
SIAL530603-10GEA
448
EUR
SIAL530603-10G
SIAL530603-1G
SIAL530603-5G
2,5-Diaminopyridine, Sigma-Aldrich®
2,5-Diaminopyridine
Formula:
C₅H₇N₃ Boiling Pt: 329 °C (1013 hPa) Melting Pt: 110.3 °C Density: 1.251 g/cm³ (20 °C) |
MDL Number:
MFCD00129023 CAS Number: 4318-76-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...