2,6-Diaminopyridine, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
D24404-100G D24404-25G D24404-500G
SIALD24404-100GEA 107 EUR
SIALD24404-100G SIALD24404-25G SIALD24404-500G
2,6-Diaminopyridine, Sigma-Aldrich®
2,6-Diaminopyridine
Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 115…118 °C
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00006329
CAS Number: 141-86-6
EINECS: 205-507-2
UN: 2671
ADR: 6.1,II