2-Hydroxypyridine ≥97%, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Pyridin-2-one, Pyridin-2(1H)-one, α-Hydroxypyridine, 2-Pyridone, α-Pyridone
M-Clarity™ quality level = MQ200
H56800-100G H56800-25G
SIALH56800-100GEA 109 EUR
SIALH56800-100G SIALH56800-25G
2-Hydroxypyridine ≥97%, Sigma-Aldrich®
2-Hydroxypyridine
Formula: C₅H₅NO
MW: 95.10 g/mol
Boiling Pt: 324 °C (1013 hPa)
Melting Pt: 107 °C
Density: 1.085 g/cm³ (30 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006268
CAS Number: 142-08-5
EINECS: 205-520-3
UN: 2811
ADR: 6.1,III