3-Pyridone 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
121851000
121855000
ACRO121851000EA
62
EUR
ACRO121851000
ACRO121855000
3-Pyridone 98%
3-Pyridone
Formula:
C₅H₅NO Boiling Pt: 180 °C (22 mmHg) Melting Pt: 123…130 °C Density: 1,2 g/cm³ (-70 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006378 CAS Number: 109-00-2 EINECS: 203-637-4 UN: 2811 |
Specification Test Results
Appearance | Beige to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 123 to 130 °C |
Water | ≤0.2 % |
GC | ≥97.5 % |
Titration with HClO4 | ≥97.5 % |