3-Pyridone, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3-Hydroxypyridine, β-Pyridone, Pyridin-3-ol, 3-Pyridinol
M-Clarity™ quality level = MQ200
H57009-100G H57009-25G H57009-500G H57009-5KG
SIALH57009-100GEA 45.6 EUR
SIALH57009-100G SIALH57009-25G SIALH57009-500G SIALH57009-5KG
3-Pyridone, Sigma-Aldrich®
3-Pyridone
Formula: C₅H₅NO
MW: 95.1 g/mol
Boiling Pt: 319
Melting Pt: 124…126 °C
Density: 1.2 g/cm³ (–70 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006378
CAS Number: 109-00-2
EINECS: 203-637-4