4-Pyridone, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 4-Hydroxypyridine, γ-Pyridone, 4-Pyridinol, Pyridin-4-ol, 1,4-Dihydro-4-Pyridinone
M-Clarity™ quality level = MQ200
120618-100G 120618-25G 120618-500G
SIAL120618-100GEA 160 EUR
SIAL120618-100G SIAL120618-25G SIAL120618-500G
4-Pyridone, Sigma-Aldrich®
4-Pyridone
Formula: C₅H₅NO
MW: 95.1 g/mol
Boiling Pt: 381
Melting Pt: 146…148 °C
MDL Number: MFCD00006419
CAS Number: 626-64-2
EINECS: 210-958-3