2-Pyridinethiol, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Pyridine-2-thiol, 2-Mercaptopyridine
M-Clarity™ quality level = MQ200
M5852-25G
M5852-5G
SIALM5852-25GEA
84.1
EUR
SIALM5852-25G
SIALM5852-5G
2-Pyridinethiol, Sigma-Aldrich®
2-Pyridinethiol
Formula:
C₅H₅NS MW: 111.17 g/mol Boiling Pt: 259 °C (1013 hPa) Melting Pt: 132.5…133.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006285 CAS Number: 2637-34-5 EINECS: 220-131-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...