4-Pyridinethiol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Pyridine-4-thiol, 4-Mercaptopyridine
M-Clarity™ quality level = MQ100
148202-10G 148202-1G
SIAL148202-10GEA 175 EUR
SIAL148202-10G SIAL148202-1G
4-Pyridinethiol, Sigma-Aldrich®
4-Pyridinethiol
Formula: C₅H₅NS
MW: 111.17 g/mol
Melting Pt: 186 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006422
CAS Number: 4556-23-4
EINECS: 224-926-1