2-Bromo-5-methylpyridine, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Bromo-5-picoline
M-Clarity™ quality level = MQ100
263354-25G 263354-5G
SIAL263354-25GEA 358 EUR
SIAL263354-25G SIAL263354-5G
2-Bromo-5-methylpyridine, Sigma-Aldrich®
2-Bromo-5-methylpyridine
Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 65 °C (0.5 mmHg)
Melting Pt: 152.5
MDL Number: MFCD00209553
CAS Number: 3510-66-5