2-Amino-5-bromo-3-methylpyridine

Supplier: Thermo Scientific
Warning

Synonyms: 2-Amino-5-bromo-3-picoline, 5-Bromo-3-methylpyridin-2-amine, 5-Bromo-3-methyl-2-pyridylamine

XBX00112.1 XBX00112EA XBX00112.50
MAYBXBX00112.1EA 0 EUR
MAYBXBX00112.1 MAYBXBX00112EA MAYBXBX00112.50
2-Amino-5-bromo-3-methylpyridine
2-Amino-5-bromo-3-methylpyridine
Formula: C₆H₇BrN₂
MW: 187.04 g/mol
MDL Number: MFCD00068232
CAS Number: 3430-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR