2-Chloro-6-methylpyridine, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 6-Chloro-2-picoline
116335-25G 116335-5G
SIAL116335-25GEA 66.9 EUR
SIAL116335-25G SIAL116335-5G
2-Chloro-6-methylpyridine, Sigma-Aldrich®
2-Chloro-6-methylpyridine
Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 64 °C (1013 hPa)
Density: 1.1506 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006245
CAS Number: 18368-63-3
EINECS: 242-241-6
UN: 2810
ADR: 6.1,III