3-Acetylpyridine, Sigma-Aldrich®
Supplier: Merck
Synonyms: Methyl 3-pyridyl ketone, 1-(Pyridin-3-yl)ethanone
M-Clarity™ quality level = MQ100
A21207-100G A21207-10G
SIALA21207-100GEA 61.5 EUR
SIALA21207-100G SIALA21207-10G
3-Acetylpyridine, Sigma-Aldrich®
3-Acetylpyridine
Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 220
Density: 1.1065 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006396
CAS Number: 350-03-8
EINECS: 206-496-7
UN: 2810
ADR: 6.1 III
Merck Index: 12,06195