2-Amino-6-methylpyridine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 6-Methyl-2-pyridylamine, 6-Methylpyridin-2-amine, 2-Amino-6-picoline
A75706-100G A75706-500G
SIALA75706-100GEA 54.6 EUR
SIALA75706-100G SIALA75706-500G
2-Amino-6-methylpyridine, Sigma-Aldrich®
2-Amino-6-methylpyridine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 214.4
Melting Pt: 40…43 °C
Storage Temperature: Ambient
MDL Number: MFCD00006331
CAS Number: 1824-81-3
EINECS: 217-360-1
UN: 2811
ADR: 6.1 II