3-Amino-2-methylpyridine, Sigma-Aldrich®
Supplier: Merck
Synonyms:
2-Methylpyridin-3-amine, 2-Methyl-3-pyridylamine, 3-Amino-2-picoline
M-Clarity™ quality level = MQ100
662690-1G
662690-250MG
SIAL662690-1GEA
206
EUR
SIAL662690-1G
SIAL662690-250MG
3-Amino-2-methylpyridine, Sigma-Aldrich®
3-Amino-2-methylpyridine
Formula:
C₆H₈N₂ MW: 108.14 g/mol |
MDL Number:
MFCD03788195 CAS Number: 3430-10-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...