6-Methyl-2-pyridone, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Hydroxy-6-methylpyridine, 6-Methylpyridin-2-ol, 6-Methylpyridin-2(1H)-one
M-Clarity™ quality level = MQ100
128740-25G 128740-5G
SIAL128740-25GEA 109 EUR
SIAL128740-25G SIAL128740-5G
6-Methyl-2-pyridone, Sigma-Aldrich®
6-Methyl-2-pyridone
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 283…284 °C (1013 hPa)
Melting Pt: 159…160 °C
Density: 1.13 g/cm³ (20 °C)
MDL Number: MFCD00006279
CAS Number: 3279-76-3
EINECS: 221-919-5