3-Ethylpyridine, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ300

W339407-100G-K W339407-1KG-K W339407-5KG-K
SIALW339407-100G-KEA 206 EUR
SIALW339407-100G-K SIALW339407-1KG-K SIALW339407-5KG-K
3-Ethylpyridine, Sigma-Aldrich®
3-Ethylpyridine
Formula: C₇H₉N
MW: 107.16 g/mol
MDL Number: MFCD00006413
CAS Number: 536-78-7
EINECS: 208-647-2
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR