3-Picoline, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
β-Picoline, 3-Methylpyridine
M-Clarity™ quality level = MQ300
P42053-1L
P42053-25ML
P42053-500ML
SIALP42053-1LEA
83.4
EUR
SIALP42053-1L
SIALP42053-25ML
SIALP42053-500ML
3-Picoline, Sigma-Aldrich®
3-Picoline
Formula:
C₆H₇N MW: 93.13 g/mol Boiling Pt: 144 °C (1013 hPa) Melting Pt: –19.4 °C Density: 0.957 g/cm³ (20 °C) Flash Pt: 38 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006402 CAS Number: 108-99-6 EINECS: 203-636-9 UN: 2313 ADR: 3,III Merck Index: 13,07485 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...