4-Picoline, Sigma-Aldrich®
Supplier: Merck
Synonyms:
4-Methylpyridine, γ-Picoline
M-Clarity™ quality level = MQ200
131490-1L
131490-250ML
131490-25ML
SIAL131490-1LEA
77.1
EUR
SIAL131490-1L
SIAL131490-250ML
SIAL131490-25ML
4-Picoline, Sigma-Aldrich®
4-Picoline
Formula:
C₆H₇N Boiling Pt: 144.9 °C (1013 hPa) Melting Pt: 3 °C Density: 0.9547 g/cm³ (20 °C) Flash Pt: 40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006440 CAS Number: 108-89-4 EINECS: 203-626-4 UN: 2313 ADR: 3,III Merck Index: 12,07556 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...