2,6-Lutidine, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
L3900-100ML-A L3900-2.5L-A L3900-500ML-A
SIALL3900-100ML-AEA 45.6 EUR
SIALL3900-100ML-A SIALL3900-2.5L-A SIALL3900-500ML-A
2,6-Lutidine, Sigma-Aldrich®
2,6-Lutidine
Formula: C₇H₉N
Boiling Pt: 143.7 °C (1013 hPa)
Melting Pt: –6 °C
Density: 0.9252 g/cm³ (20 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00006345
CAS Number: 108-48-5
EINECS: 203-587-3
UN: 1993
ADR: 3,III
Merck Index: 13,05636