3,5-Lutidine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 3,5-Dimethylpyridine
M-Clarity™ quality level = MQ200
L4206-100ML L4206-500ML
SIALL4206-100MLEA 78.5 EUR
SIALL4206-100ML SIALL4206-500ML
3,5-Lutidine, Sigma-Aldrich®
3,5-Lutidine
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 172.7 °C (1013 hPa)
Melting Pt: –9 °C
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00006404
CAS Number: 591-22-0
EINECS: 209-708-6
UN: 1992
ADR: 3,III