2,4,6-Collidine, Sigma-Aldrich®
Supplier: Merck
Danger
142387-100ML 142387-500ML 142387-5ML
SIAL142387-100MLEA 109 EUR
SIAL142387-100ML SIAL142387-500ML SIAL142387-5ML
2,4,6-Collidine, Sigma-Aldrich®
2,4,6-Collidine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 171
Melting Pt: –43 °C
Density: 0.9191 g/cm³ (20 °C)
Flash Pt: 55
Storage Temperature: Ambient
MDL Number: MFCD00006338
CAS Number: 108-75-8
EINECS: 203-613-3
UN: 1992
ADR: 3 III
Merck Index: 13,09789