(S)-(-)-Nicotine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: (S)-(-)-3-(N-Methyl-2-pyrrolidinyl)pyridine, (S)-(-)-3-(1-Methyl-2-pyrrolidinyl)pyridine, L(-)-Nicotine, L-Nicotine, Nicotine, (-)-Nicotine
M-Clarity™ quality level = MQ200
N3876-25ML N3876-100ML N3876-5ML
SIALN3876-25MLEA 309 EUR
SIALN3876-25ML SIALN3876-100ML SIALN3876-5ML
(S)-(-)-Nicotine, Sigma-Aldrich®
(S)-(-)-Nicotine
Formula: C₁₀H₁₄N₂
MW: 162.23 g/mol
Boiling Pt: 247
Melting Pt: –79 °C
Density: 1.01 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00006369
CAS Number: 54-11-5
EINECS: 200-193-3
UN: 1654
ADR: 6.1 II
Merck Index: 12,06611