Benzyl nicotinate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3-Picolinic acid benzyl ester, Pyridine-3-carboxylic acid benzyl ester, Benzyl 3-pyridinecarboxylate, 3-Pyridinecarboxylic acid benzyl ester, Nicotinic acid benzyl ester
72351-100ML 72351-500ML
SIAL72351-100MLEA 94.6 EUR
SIAL72351-100ML SIAL72351-500ML
Benzyl nicotinate, Sigma-Aldrich®
Benzyl nicotinate
Formula: C₁₃H₁₁NO₂
MW: 213.24 g/mol
Boiling Pt: 179…180 °C (8 hPa)
Melting Pt: 24
Density: 1.17 g/cm³ (20 °C)
MDL Number: MFCD00023584
CAS Number: 94-44-0
EINECS: 202-332-3