2-Phenylpyridine, Sigma-Aldrich®
Supplier: Merck
Warning
P33402-10G P33402-50G
SIALP33402-10GEA 90.6 EUR
SIALP33402-10G SIALP33402-50G
2-Phenylpyridine, Sigma-Aldrich®
2-Phenylpyridine
Formula: C₁₁H₉N
MW: 155.2 g/mol
Boiling Pt: 269…270 °C (1013 hPa)
Melting Pt: 76…77 °C
Density: 1.623 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006280
CAS Number: 1008-89-5
EINECS: 213-763-1
UN: 2810
ADR: 6.1 III