2-Bromopyrimidine, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

245844-1G 245844-5G
SIAL245844-1GEA 27.2 EUR
SIAL245844-1G SIAL245844-5G
2-Bromopyrimidine, Sigma-Aldrich®
2-Bromopyrimidine
Formula: C₄H₃BrN₂
MW: 158.99 g/mol
Boiling Pt: 62…64 °C (1.5 mmHg)
Melting Pt: 56…57.5 °C
Density: >1 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00014601
CAS Number: 4595-60-2
EINECS: 224-993-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR