2,4,5-Trichloropyrimidine, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
652032-1G
652032-5G
SIAL652032-1GEA
57.3
EUR
SIAL652032-1G
SIAL652032-5G
2,4,5-Trichloropyrimidine, Sigma-Aldrich®
2,4,5-Trichloropyrimidine
Formula:
C₄HCl₃N₂ MW: 183.42 g/mol Boiling Pt: 94 °C (12 mmHg) |
MDL Number:
MFCD03788200 CAS Number: 5750-76-5 UN: 3334 ADR: 9, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...