2,4-Dichloropyrimidine, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
143847-10G 143847-50G
SIAL143847-10GEA 79.5 EUR
SIAL143847-10G SIAL143847-50G
2,4-Dichloropyrimidine, Sigma-Aldrich®
2,4-Dichloropyrimidine
Formula: C₄H₂Cl₂N₂
MW: 148.98 g/mol
Boiling Pt: 209
Melting Pt: 61
MDL Number: MFCD00006061
CAS Number: 3934-20-1
EINECS: 223-508-6