2-Chloropyrimidine, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
193291-10G
193291-50G
SIAL193291-10GEA
59.1
EUR
SIAL193291-10G
SIAL193291-50G
2-Chloropyrimidine, Sigma-Aldrich®
2-Chloropyrimidine
Formula:
C₄H₃ClN₂ Boiling Pt: 199 °C (1013 hPa) Melting Pt: 65…66 °C |
MDL Number:
MFCD00006060 CAS Number: 1722-12-9 EINECS: 217-020-2 UN: 2811 ADR: 6.1,III |