2,4-Difluoropyrimidine, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
661082-100MG
661082-500MG
SIAL661082-100MGEA
49.7
EUR
SIAL661082-100MG
SIAL661082-500MG
2,4-Difluoropyrimidine, Sigma-Aldrich®
2,4-Difluoropyrimidine
Formula:
C₄H₂F₂N₂ MW: 116.07 g/mol |
MDL Number:
MFCD04972830 CAS Number: 2802-61-1 UN: 2924 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...