Dihydropyrimidine-2,4,5(3H)-trione ≥98%
Supplier: Thermo Scientific
Warning
Synonyms: Isobarbitursäure, Isobarbituric acid, 2,4,5-Trihydroxypyrimidine
B23320.03 B23320.06
B23320.03EA 67.8 EUR
B23320.03 B23320.06
Dihydropyrimidine-2,4,5(3H)-trione ≥98%
Dihydropyrimidine-2,4,5(3H)-trione
Purity: 98% CAS No.: 496-76-4 Molecular Formula: C4H4N2O3 Formula Weight: 128.09 Melting Point: 300° Beilstein Reference: 127205
Formula: C₄H₄N₂O₃
MW: 128.09 g/mol
Boiling Pt: 410 °C (1013 hPa)
Melting Pt: 330…340 °C
MDL Number: MFCD00082987
CAS Number: 496-76-4
EINECS: 207-829-9