2,4-Dichloro-6-methylpyrimidine ≥98%
Supplier: Thermo Fisher Scientific
Danger
A10363.14 A10363.22
A10363.14EA 111 EUR
A10363.14 A10363.22
2,4-Dichloro-6-methylpyrimidine ≥98%
2,4-Dichloro-6-methylpyrimidine
Purity: 98% CAS No.: 5424-21-5 Molecular Formula: C5H4Cl2N2 Formula Weight: 163.01 Flash Point: 110°(230°F) Melting Point: 44-47° Boiling Point: 219° Beilstein Reference: 114295 UN No.: 1759 MDL No.: MFCD00006064
Formula: C₅H₄Cl₂N₂
Boiling Pt: 219 °C
Melting Pt: 44…47 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006064
CAS Number: 5424-21-5
EINECS: 226-563-4
UN: 1759
ADR: 8,II