Pyrrole, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 1H-Pyrrole

M-Clarity™ quality level = MQ200

131709-100ML 131709-25ML 131709-500ML
SIAL131709-100MLEA 89.8 EUR
SIAL131709-100ML SIAL131709-25ML SIAL131709-500ML
Pyrrole, Sigma-Aldrich®
Pyrrole
Formula: C₄H₅N
Boiling Pt: 130…131 °C (1013 hPa)
Melting Pt: –24 °C
Density: 0.97 g/cm³ (20 °C)
Flash Pt: 39 °C
MDL Number: MFCD00005216
CAS Number: 109-97-7
EINECS: 203-724-7
UN: 1992
ADR: 3,III
Merck Index: 12,08198

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR