Pyrrole, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1H-Pyrrole
M-Clarity™ quality level = MQ300
W338605-100G W338605-1KG W338605-5KG W338605-SAMPLE
SIALW338605-100GEA 64.5 EUR
SIALW338605-100G SIALW338605-1KG SIALW338605-5KG SIALW338605-SAMPLE
Pyrrole, Sigma-Aldrich®
Pyrrole
Formula: C₄H₅N
MW: 67.09 g/mol
Boiling Pt: 130…131 °C (1013 hPa)
Melting Pt: –24 °C
Density: 0.97 g/cm³ (20 °C)
Flash Pt: 39
MDL Number: MFCD00005216
CAS Number: 109-97-7
EINECS: 203-724-7
UN: 1992
ADR: 3 III
Merck Index: 12,08198