Octamethyltrisiloxane ≥98%, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Dimethylbis(trimethylsilyloxy)silane, Poly(dimethylsiloxane), 1,1,1,3,3,5,5,5-Octamethyltrisiloxane

M-Clarity™ quality level = MQ100

235709-100ML 235709-5ML
SIAL235709-100MLEA 246 EUR
SIAL235709-100ML SIAL235709-5ML
Octamethyltrisiloxane ≥98%, Sigma-Aldrich®
Octamethyltrisiloxane, kinematic viscosity 1.0 cSt (25°C)
Formula: [(CH₃)₃SiO]₂Si(CH₃)₂
MW: 236.53 g/mol
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: –82 °C
Density: 0.82 g/cm³ (20 °C)
Flash Pt: 29 °C (closed cup)
MDL Number: MFCD00008264
CAS Number: 107-51-7
EINECS: 203-497-4
UN: 1993
ADR: 3,III
Merck Index: 12,06844
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fba031046b7091a87c7b3f3b170c026c07722977&vid=850e79089a70db712f48c959a31f0c51041e5bfe&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR