3,4-Dibromothiophene, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
247154-25G 247154-5G
SIAL247154-25GEA 236 EUR
SIAL247154-25G SIAL247154-5G
3,4-Dibromothiophene, Sigma-Aldrich®
3,4-Dibromothiophene
Formula: C₄H₂Br₂S
MW: 241.93 g/mol
Boiling Pt: 221 °C (1013 hPa)
Melting Pt: 4.5 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005465
CAS Number: 3141-26-2
EINECS: 221-546-8