Thiophene, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
T31801-100G T31801-500G T31801-5G
SIALT31801-100GEA 32 EUR
SIALT31801-100G SIALT31801-500G SIALT31801-5G
Thiophene, Sigma-Aldrich®
Thiophene
Formula: C₄H₄S
MW: 84.14 g/mol
Boiling Pt: 84.4 °C (1013 hPa)
Melting Pt: –38 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: –9 °C
Storage Temperature: Ambient
MDL Number: MFCD00005413
CAS Number: 110-02-1
EINECS: 203-729-4
UN: 2414
ADR: 3,II
Merck Index: 12,09490