2-Chloro-3-methylthiophene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
648760-1G
648760-5G
SIAL648760-1GEA
37.4
EUR
SIAL648760-1G
SIAL648760-5G
2-Chloro-3-methylthiophene, Sigma-Aldrich®
2-Chloro-3-methylthiophene
Formula:
C₅H₅ClS MW: 132.61 g/mol Boiling Pt: 153…157 °C (1013 hPa) Density: 1.2281 g/cm³ (25 °C) |
MDL Number:
MFCD00130083 CAS Number: 14345-97-2 EINECS: 238-296-0 |
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...