2-Methylthiophene, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ100
M84208-100G
M84208-25G
SIALM84208-100GEA
106
EUR
SIALM84208-100G
SIALM84208-25G
2-Methylthiophene, Sigma-Aldrich®
2-Methylthiophene
Formula:
C₅H₆S MW: 98.17 g/mol Boiling Pt: 113 °C (1013 hPa) Melting Pt: –63 °C Density: 1.02 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005451 CAS Number: 554-14-3 EINECS: 209-063-0 UN: 1993 ADR: 3,II |
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...